Learn More
Tetraethyl (dimethylaminomethylene)diphosphonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Tetraethyl (dimethylaminomethylene)diphosphonate undergoes Horner-Wadsworth-Emmons olefination with an aldehyde gives the ?-phosphonato enamine, readily convertible to the homologous acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetraethyl (dimethylaminomethylene)diphosphonate undergoes Horner-Wadsworth-Emmons olefination with an aldehyde gives the ɑ-phosphonato enamine, readily convertible to the homologous acid.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Tetraethyl (dimethylaminomethylene)diphosphonate undergoes Horner-Wadsworth-Emmons olefination with an aldehyde gives the ɑ-phosphonato enamine, readily convertible to the homologous acid.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Specifications
Boiling Point | 138°C to 140°C (1 mmHg) |
Refractive Index | 1.451 |
Quantity | 5 g |
Beilstein | 1914890 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 331.29 |
Percent Purity | 97% |
Chemical Name or Material | Tetraethyl (dimethylaminomethylene)diphosphonate |