missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl nitrite, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7058448
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
540-80-7
C4H9NO2
103.121
MFCD00002055
IOGXOCVLYRDXLW-UHFFFAOYSA-N
t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
10906
tert-butyl nitrite
CC(C)(C)ON=O
This item is not returnable. View return policy
540-80-7
C4H9NO2
103.121
MFCD00002055
IOGXOCVLYRDXLW-UHFFFAOYSA-N
t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
10906
tert-butyl nitrite
CC(C)(C)ON=O

tert-Butyl nitrite is employed as a reagent for diazotization and nitrosation of alcohols, thiols, amines and cycloalkanes. It plays an important role in photocatalyzed conversion of aryl and heteroarylamines to selenides. It finds application in the preparation of aryl azides from aryl amines. It is also useful for radical multifunctionalization reactions of aliphatic alkenes and source of nitric oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl nitrite is employed as a reagent for diazotization and nitrosation of alcohols, thiols, amines and cycloalkanes. It plays an important role in photocatalyzed conversion of aryl and heteroarylamines to selenides. It finds application in the preparation of aryl azides from aryl amines. It is also useful for radical multifunctionalization reactions of aliphatic alkenes and source of nitric oxide.

Solubility
Miscible with diethyl ether, chloroform, alcohol and carbon disulfide. Slightly miscible with water. Immiscible with glycerol.

Notes
Light sensitive. Store in cool place. Incompatible with strong reducing agents, powdered metals and strong acids.
TRUSTED_SUSTAINABILITY
Density 0.867
Boiling Point 61°C to 63°C
Flash Point −23°C (−9°F)
Odor Sweet
Refractive Index 1.369
Quantity 25 g
UN Number UN2351
Beilstein 1209339
Merck Index 14,1583
Solubility Information Miscible with diethyl ether,chloroform,alcohol and carbon disulfide. Slightly miscible with water. Immiscible with glycerol.
Formula Weight 103.12
Percent Purity 90%
Grade Technical
Chemical Name or Material tert-Butyl nitrite
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'