Learn More
tert-Butyl ethyl malonate, 97%, Thermo Scientific Chemicals
Catalog Number p-3770847
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
32864-38-3
C9H16O4
188.22
MFCD00009193
OCOBFMZGRJOSOU-UHFFFAOYSA-N
tert-butyl ethyl malonate, t-butyl ethyl malonate, propanedioic acid, 1,1-dimethylethyl ethyl ester, 1-tert-butyl 3-ethyl propanedioate, tert-butyl ethyl propanedioate, malonic acid tert-butyl ethyl ester, tert-butyl ethyl propane-1,3-dioate, zlchem 692, acmc-1cjju, 1-tert-butyl ethyl malonate
96345
3-O-tert-butyl 1-O-ethyl propanedioate
CCOC(=O)CC(=O)OC(C)(C)C
32864-38-3
C9H16O4
188.22
MFCD00009193
OCOBFMZGRJOSOU-UHFFFAOYSA-N
tert-butyl ethyl malonate, t-butyl ethyl malonate, propanedioic acid, 1,1-dimethylethyl ethyl ester, 1-tert-butyl 3-ethyl propanedioate, tert-butyl ethyl propanedioate, malonic acid tert-butyl ethyl ester, tert-butyl ethyl propane-1,3-dioate, zlchem 692, acmc-1cjju, 1-tert-butyl ethyl malonate
96345
3-O-tert-butyl 1-O-ethyl propanedioate
CCOC(=O)CC(=O)OC(C)(C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.0010g/mL
Boiling Point 106.0°C to 110.0°C
Flash Point 87°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CO2CCH2CO2C2H5
Refractive Index 1.4163
Quantity 1 g
Specific Gravity 1.001
Solubility Information Solubility in water: insoluble (20°C). Other solubilities: soluble in most common solvents
Formula Weight 188.22
Percent Purity 97%
Chemical Name or Material tert-Butyl ethyl malonate