Learn More
tert-Butyl 4-(aminocarbothioyl)tetrahydropyridine-1(2H)-carboxylate, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 244.36 |
Percent Purity | 95% |
Chemical Name or Material | tert-Butyl 4-(aminocarbothioyl)tetrahydropyridine-1(2H)-carboxylate |