Learn More
tert-Butyl 4-aminobutyrate hydrochloride, 97%, Thermo Scientific Chemicals
614.43 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Employed as an important intermediate for raw material for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as an important intermediate for raw material for pharmaceutical.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Employed as an important intermediate for raw material for pharmaceutical.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Color | White |
Linear Formula | H2NCH2CH2CH2CO2C(CH3)3·HCl |
Formula Weight | 195.69 |
Percent Purity | 97% |
Quantity | 1 g |
Physical Form | Powder |
Chemical Name or Material | tert-Butyl 4-aminobutyrate hydrochloride |