missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[5-(ethoxycarbonyl)-4-methyl -1,3-thiazol-2-yl]tetrahydro-1(2H)-pyridine, ≥90%, Thermo Scientific™

Catalog Number 10596123
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 852180-50-8
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C17H26N2O4S
MDL Number MFCD07368516
Quantity 1 g
Synonym tert-butyl 4-5-ethoxycarbonyl-4-methyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridinecarboxylate, tert-butyl 4-5-ethoxycarbonyl-4-methyl-1,3-thiazol-2-yl piperidine-1-carboxylate, 4-5-ethoxycarbonyl-4-methyl-1,3-thiazol-2-yl piperidine, n-boc protected, ethyl 4-methyl-2-n-boc-piperidin-4-yl thiazol-5-yl carboxylate, ethyl 2-1-tert-butoxycarbonyl piperidin-4-yl-4-methylthiazole-5-carboxylate, tert-butyl 4-5-ethoxycarbonyl-4-methyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridine, 1-piperidinecarboxylicacid, 4-4-methyl-5-1-oxopropyl-2-thiazolyl-, 1,1-dimethylethyl ester, ethyl 4-methyl-2-1-2-methylpropan-2-yl oxycarbonyl piperidin-4-yl-1,3-thiazole-5-carboxylate
InChI Key YTFTXLAXZXXHPI-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(N=C(S1)C2CCN(CC2)C(=O)OC(C)(C)C)C
IUPAC Name ethyl 4-methyl-2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol) 354.465
PubChem CID 7060597
Formula Weight 354.47
Percent Purity 90%
Chemical Name or Material tert-Butyl 4-[5-(ethoxycarbonyl)-4-methyl-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'