Learn More
tert-Butyl (3R)-3-amino-5-methylhexanoate, 97%, Thermo Scientific™
Catalog Number p-81187
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
166023-29-6
C11H24NO2
202.32
MFCD01076257
WBOPVTYXLFEVGP-SECBINFHSA-O
2733825
tert-butyl (3R)-3-amino-5-methylhexanoate
CC(C)C[C@@H]([NH3+])CC(=O)OC(C)(C)C
166023-29-6
C11H24NO2
202.32
MFCD01076257
WBOPVTYXLFEVGP-SECBINFHSA-O
2733825
tert-butyl (3R)-3-amino-5-methylhexanoate
CC(C)C[C@@H]([NH3+])CC(=O)OC(C)(C)C
Color Colorless to Yellow
Assay Percent Range 96% min. (GC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Synonym tert-butyl 3r-3-amino-5-methylhexanoate, hexanoicacid, 3-amino-5-methyl-, 1,1-dimethylethyl ester, 3r, tert-butyl 3r-3-amino-5-methyl-hexanoate, r-tert-butyl 3-amino-5-methylhexanoate, 3r-3-amino-5-methylhexanoic acid tert-butyl ester, hexanoicacid,3-amino-5-methyl-,1,1-dimethylethyl ester, 3r
Refractive Index 1.4300 to 1.432
Specific Rotation -15
Formula Weight 201.31
Percent Purity 97%
Quantity 100 mg
Physical Form Liquid
Chemical Name or Material tert-Butyl (3R)-3-amino-5-methylhexanoate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (3R)-3-amino-5-methylhexanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.