missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanol, TEBOL™, ≥99.3%, Honeywell™

Catalog Number p-7120860
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
This item is not returnable. View return policy
75-65-0
C4H10O
74.123
MFCD00004464
DKGAVHZHDRPRBM-UHFFFAOYSA-N
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O
This item is not returnable. View return policy
75-65-0
C4H10O
74.123
MFCD00004464
DKGAVHZHDRPRBM-UHFFFAOYSA-N
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O

TEBOL® 99,≥99.3%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Melting Point 26°C
Color Colorless
Linear Formula (CH3)3COH
UN Number UN1120
Beilstein 906698
Formula Weight 74.12g/mol
Percent Purity ≥99.3%
Density 0.775g/mL at 25°C
Boiling Point 83°C
Quantity 500 mL
Explosion Limit 0.08
Flash Point 11°C (51.8°F)
Ignition Point 896°F
Odor Characteristic
Packaging Poly Bottle
Physical Form Liquid
Vapor Pressure 31mmHg (20°C), 44mmHg (26°C)
Chemical Name or Material tert-Butanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'