missing translation for 'onlineSavingsMsg'
Learn More

TER-119, APC-eFluor 780, clone: TER-119, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 47-5921-82

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15371660

  • 1638.00 NOK / 100µg


In Stock 7
Add to basket

Description

Description

Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin.Applications Reported: This TER-119 antibody has been reported for use in flow cytometric analysis.Applications Tested: This TER-119 antibody has been tested by flow cytometric analysis of mouse bone marrow. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.APC-eFluor™ emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TER-119
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
Liquid
Ly76
IgG2b, kappa, kappa
100μg
4° C, store in dark, DO NOT FREEZE!, handling and experimental procedures.
Primary
104231
Flow Cytometry
TER-119
APC-eFluor 780
Ly76
TER119, Erythroid cell marker, Ly-76, Ly76
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Videos

For Research Use Only.