Learn More
Fisher Chemical 125 ML Ten elements solution - Ca (500mg/L), Co (20mg/L), Cu (20mg/L), Fe (20mg/L), Mg (25mg/L), (Zn 20mg/L) for ICP/MS 2% HNO3 matrix 125mL
Brand: Fisher Chemical CM1020
125 ML Ten elements solution - Ca (500mg/L), Co (20mg/L), Cu (20mg/L), Fe (20mg/L), Mg (25mg/L), (Zn 20mg/L) for ICP/MS 2% HNO3 matrix 125mL
Additional Details : Weight : 0.12500kg