Learn More
Tariquidar methanesulfonate, hydrate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Tariquidar methanesulfonate, hydrate (XR9576 methanesulfonate, hydrate) is a potent and specific inhibitor of P-glycoprotein (P-gp) with a Kd of 5.1 nM.
Specifications
Specifications
| Chemical Name or Material | Tariquidar methanesulfonate, hydrate |
| CAS | 625375-83-9 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C40H52N4O15S2 |
| Quantity | 10 mg |
| Synonym | XR9576 methanesulfonate, hydrate |
| Solubility Information | DMSO : ≥ 296 mg/mL (331.47 mM) ∣H2O : 5 mg/mL (5.60 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC2=CC=CC=C2N=C1)NC3=CC(OC)=C(OC)C=C3C(NC4=CC=C(CCN5CC6=C(C=C(OC)C(OC)=C6)CC5)C=C4)=O.CS(=O)(O)=O.CS(=O)(O)=O.O.O.O |
| Molecular Weight (g/mol) | 892.99 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.