Learn More
Applied Biosystems™ TaqMan™ Array, Human Colorectal Cancer Metastasis, Fast 96-well
Product Code. 10014934 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 96-well plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10014934 1 x 96-well plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10014934 Supplier Applied Biosystems™ Supplier No. 4418766
  • ONLINE EXCLUSIVE PRICE
    2215.00 NOK
    1971.35 NOK / 1 sets
    Save 243.65 NOK
    11% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.
  • These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state
  • Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment
  • The TaqMan™ Array Human Colorectal Cancer Metastasis 96-well Plate contains:
    • 92 assays to colorectal cancer metastasis associated genes
    • 4 assays to candidate endogenous control genes
TRUSTED_SUSTAINABILITY
Specifications
Type Array Plate
Format 96-array Plate
Species Human
Content And Storage Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.
Form Dried Down
Includes TaqMan Assay for One 10 uL Reaction
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 1 x 96-well plate
Reaction Speed Fast
Research Category Neurobiology, Biomarker-Related Pathway(s), Development, Oncology