missing translation for 'onlineSavingsMsg'
Learn More

Syntaxin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15556-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884784

  • 3468.15 NOK / 150µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The gene is a member of the syntaxin family. The encoded protein is targeted to the apical membrane of epithelial cells where it forms clusters and is important in establishing and maintaining polarity necessary for protein trafficking involving vesicle fusion and exocytosis. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

Syntaxin 3
Polyclonal
Unconjugated
STX3
STX3, STX3A, syntaxin 3
Rabbit
Antigen Affinity Chromatography
RUO
20908, 6809, 81802
-20°C
Liquid
Western Blot, Immunoprecipitation
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q08849, Q13277, Q64704
Stx3
Syntaxin 3 Fusion Protein Ag7925
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers