Learn More
Hettich™ Swing-out Rotor for ROTINA 380/380 R Centrifuge, 2-place
Provides consistent, accurate results for blood, cell/tissue culture, plates or bottle applications. The ROTINA 380 R is a high capacity, general purpose, benchtop centrifuge with a small footprint.
Brand:  Hettich™ 1760
Additional Details : Weight : 0.80000kg
Product Code. 10236992
  • 10575.00 NOK / Each
Estimated Shipment: 07-05-2024
Description
  • For use with ROTINA 380/380 R
  • The ROTINA 380's versatility and size make an invaluable addition to any lab. Ideal for clinical, research, food and beverage and industrial applications.
  • Robust and steady high-capacity centrifuge
  • Accessories with bio-containment protect users from aerosols while ensuring optimal safety, for instance, while centrifuging infectious samples
  • Temperature control system with pre-cool function from -20 to +40°C
  • Versa-Rotor™ for multiple applications
  • All-steel construction
  • Quiet Operation
  • Easy-Lift™/Safety-Lock™ Lid
Specifications
Specifications
2,397 x g
134 mm
90°
4,000 rpm
ROTINA 380/380 R
2