missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Antibody (60.11) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB500-238JF669
This item is not returnable.
View return policy
Description
Survivin Monoclonal antibody specifically detects Survivin in Human,Mouse,Rat samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Proximity Ligation Assay,Flow (Intracellular),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| Survivin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| This Survivin Antibody (60.11) [Alexa Fluor 350] was developed against full length recombinant human Survivin [UniProt# O15392] | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
| 332 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Proximity Ligation Assay, Flow Cytometry (Intracellular Staining), KnockDown, Immunocytochemistry/Immunofluorescence | |
| 60.11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction