Learn More
Uvex™ Super Boss Safety Helmet
A versatile work safety helmet that allows very good climate control thanks to its adjustable side ventilation slits.
Brand:  Uvex™ 9750120
Additional Details : Weight : 8.25000kg
 View more versions of this product
Product Code. 17594076
  • 2365.00 NOK / Pack of 25
Estimated Shipment: 06-11-2023
to see stock.
Add to basket
Description
Description
  • Size and weight optimized helmet
  • Lateral adjustable air vents
  • 6-point textile suspension harness with sweatband
  • Side slots for attaching ear defenders or visors
Specifications
Specifications
EN 397
HDPE
Industrial Safety Helmet
Ventilated
Yellow
9750
Long Brim