Learn More
Uvex™ Super Boss Safety Helmet
A versatile work safety helmet that allows very good climate control thanks to its adjustable side ventilation slits.
Brand:  Uvex™ 9750020
1576.67 NOK valid until 2023-05-26
Use promo code "20831" to get your promotional price.
Additional Details : Weight : 8.25000kg
 View more versions of this product
Product Code. 17584076
  • 2365.00 NOK / Pack of 25
Estimated Shipment: 13-07-2023
Description
  • Size and weight optimized helmet
  • Lateral adjustable air vents
  • 6-point textile suspension harness with sweatband
  • Side slots for attaching ear defenders or visors
Specifications
Specifications
EN 397
HDPE
Industrial Safety Helmet
Yes
White
9750
Long Brim