Learn More
Uvex™ Super Boss Safety Helmet
A versatile work safety helmet that allows very good climate control thanks to its adjustable side ventilation slits.
Brand: Uvex™ 9750020
1576.67 NOK valid until 2023-05-26
Use promo code "20831" to get your promotional price.
Additional Details : Weight : 8.25000kg
Description
- Size and weight optimized helmet
- Lateral adjustable air vents
- 6-point textile suspension harness with sweatband
- Side slots for attaching ear defenders or visors
Specifications
EN 397 | |
HDPE | |
Industrial Safety Helmet | |
Yes |
White | |
9750 | |
Long Brim |