Learn More
STXBP1 Mouse anti-Human, Unconjugated, Clone: 1B5B3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67137-1-IG-20UL
 View more versions of this product
Product Code. 16849595
  • 1611.84 NOK / 20µL
Explore more special offers
Description
Description
The STXBP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STXBP1. This antibody recognizes human, mouse, rat, pig antigen. The STXBP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
STXBP1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence
1B5B3
Unconjugated
20 μL
6812
IgG2a