Learn More
Molecular Probes™ Streptavidin-CY3 (ZyMax Grade)
Streptavidin-CY3 (ZyMax Grade)
Brand: Molecular Probes™ 438315
2166.77 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Applications: Immunohistochemistry, Western blotting, ELISA, ISHZyMAX™ reagents offer enhanced assay sensitivity, higher titer, and greater flexibility in assay design than standard grade reagents.
Specifications
1 mg/mL | |
Store in refrigerator (2–8°C). | |
Liquid | |
Streptavidin Conjugate (fluorescent) |
Cy3 | |
Wet Ice | |
Invitrogen™ |
For Research Use Only. Not for use in diagnostic procedures.