Learn More
Thermo Scientific Chemicals Standard Sample Tube, 3.24mm ID
For use with MKII Magnetic Susceptibility Balances
Brand:  Thermo Scientific Chemicals 014349.KL
10567.19 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11480400
  • 16646.40 NOK / Pack of 20
Estimated Shipment: 30-04-2024
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • I.D.: 3.24mm
Specifications
Specifications
20 Pack
Standard Sample Tube