Learn More
Tenak™ Standard Rack for Upright Freezer, 50 mm Box Height, with Boxes
Shop All TENAK Products
Click to view available options
Holds:
Boxes
Array:
3 x 3
3 x 4
3 x 5
4 x 3
4 x 5
5 x 3
5 x 4
5 x 5
6 x 4
6 x 5
7 x 4
Unit Size:
Each
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Spécification
Spécification
| Includes | White cardboard boxes, 9 x 9 cell dividers |
| Array | 5 x 4 |
| Holds | Boxes |
| Material | Stainless Steel |
| Positions | 20 |
| For Use With (Equipment) | Upright Freezers |
| Dimensions (L x W x H) | 560 x 139 x 276 mm |
| Height (Metric) | 50 mm (Box) |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu