Learn More
VACUUBRAND™ Stainless Steel T-Pieces With Small KF Flanges
Brand:  VACUUBRAND™ 20673515
1739.20 NOK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10009600
  • 2395.00 NOK / Each
Estimated Shipment: 21-05-2024
Specifications
25/25/25 mm
Stainless Steel
50 x 100 mm
Solenoid-operated valves VV and VV C
1 Each
T-piece