Learn More
Merck Millipore Stainless Steel Funnel
Brand:  Merck Millipore XF2004756
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10025431
  • 1748.00 NOK / Each
Estimated Shipment
29-03-2021
Specifications
250mL
Stainless Steel
Funnel, 100mL, Stainless Steel
250mL