Learn More
Thermo Scientific™ Stainless-Steel Disposable Resin
Refill stainless-steel ion exchanges with this disposable, mixed-bed resin featuring advanced, regeneration service.
3760.35 NOK - 7379.95 NOK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13182116
|
Thermo Scientific™
50155540 |
Resin 2 x 22 liters disposable |
7379.95 NOK
Pack of 2 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11825912
|
Thermo Scientific™
50155538 |
Resin 2 x 12 liters disposable |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Use Thermo Scientific™ Disposable Resin to refill stainless steel ion exchangers. The mixed-bed resins have documented quality monitoring and reliably function due to an advanced regeneration service. Each regenerated batch is monitored for both consistent capacity and quality.
This service is only available in Germany.
Specifications
Stainless Steel | |
Mixed Bed | |
Disposable Resin |
This service is only available in Germany.