Learn More
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire
Adjusted to Meet or Exceed OIML E1 Tolerances
940.00 NOK
Specifications
Certifications/Compliance | No Certificate |
---|---|
Material | Stainless Steel (316) |
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16662292
|
Ohaus™
30517704 |
1 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16672292
|
Ohaus™
30517705 |
2 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16682292
|
Ohaus™
30517706 |
5 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16692292
|
Ohaus™
30517707 |
10 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16602302
|
Ohaus™
30517958 |
20 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16612302
|
Ohaus™
30517959 |
50 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16622302
|
Ohaus™
30517960 |
100 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16632302
|
Ohaus™
30517961 |
200 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16642302
|
Ohaus™
30517962 |
500 mg |
940.00 NOK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
No Certificate | |
Stainless Steel (316) | |
Protective Case |