Learn More
Schneider Gerd™ Stainless Steel 4021 Tissue Forceps
Schneider Gerd™ Stainless Steel 4021 Tissue Forceps is used to handle delicate tissues.
Brand:  Schneider Gerd™ 107
Additional Details : Weight : 0.01980kg
Product Code. 11397364
  • 331.00 NOK / Each
Estimated Shipment: 31-05-2024
Description
  • Made of Stainless steel 4021
  • Length: 130mm (5.11 in.)
Specifications
Specifications
5.11 in.
Stainless Steel (4021)
130 mm
Forceps, Tissue