missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSRP1 Mouse anti-Human, Unconjugated, Clone: 3D3H4, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | SSRP1 |
---|---|
Clone | 3D3H4 |
Concentration | 500 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889865
|
Proteintech
67313-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16879865
|
Proteintech
67313-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The SSRP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SSRP1. This antibody recognizes human, mouse, rat antigen. The SSRP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SSRP1 | |
500 μg/mL | |
Monoclonal | |
Mouse | |
6749 | |
Primary |
3D3H4 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2b |