missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRC Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | SRC |
---|---|
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887943
|
Proteintech
11097-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877943
|
Proteintech
11097-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SRC is a non-receptor kinase involved in the regulation of growth and differentiation of eukaryotic cells. SRC associates with the cytoplasmic region of various cell surface receptors to facilitate signal transduction, and is also involved in cytoskeletal organization, cell cycle regulation, and apoptosis. In humans, the gene encoding SRC is present on chromosome 20. Activity of SRC is regulated by two tyrosine residues (Tyr 416 and Tyr527), and phosphorylation of these two sites have opposing effects. Phosphorylation of Tyr416 in the activation loop of the kinase domain upregulates enzyme activity while the phosphorylation of Tyr527 decreases enzyme activity. Mutations in the SRC gene could be involved in the maligt progression of colon cancer. Two transcript variants encoding the same SRC protein have been found thus far.Specifications
SRC | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Hamster, Human, Mouse, Rat | |
P05480, P12931, Q9WUD9 | |
100757188, 20779, 6714, 83805 | |
SRC Fusion Protein Ag1575 | |
Primary | |
-20°C | |
SRC |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ASV, c SRC, p60 Src, pp60c src, Proto oncogene c Src, SRC, SRC1 | |
SRC | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |