Learn More
Uvex Sport Blue PU 65991 Low Shoe - Width 10
Offers first-class wearer comfort with a sporty design; low weight, climate-optimizes high-tech materials and ergonomic outsole
1530.00 NOK
Specifications
Color | Blue |
---|---|
Material | Textile |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17203071
|
Uvex
6599135 |
35 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17213071
|
Uvex
6599136 |
36 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17223071
|
Uvex
6599137 |
37 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17233071
|
Uvex
6599138 |
38 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17243071
|
Uvex
6599139 |
39 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17253071
|
Uvex
6599140 |
40 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17263071
|
Uvex
6599141 |
41 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17273071
|
Uvex
6599142 |
42 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17283071
|
Uvex
6599144 |
44 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17293071
|
Uvex
6599145 |
45 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17203081
|
Uvex
6599146 |
46 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17213081
|
Uvex
6599147 |
47 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17223081
|
Uvex
6599148 |
48 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17233081
|
Uvex
6599149 |
49 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17243081
|
Uvex
6599150 |
50 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17253081
|
Uvex
6599151 |
51 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17263081
|
Uvex
6599152 |
52 |
1530.00 NOK
1 pair |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
Uvex 1 Sport Shoe 6599.1 S1 P SRC
- Modern, ultra-lightweight and flexible S1 safety shoe
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials are free of silicones plasticizers and other paint wetting impairment substances
- Meets ESD specification with leakage resistance less than 35 megohms
- 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good lateral stability, no thermal conductivity
- Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility of shoe
- Made with new developed technology for exceptional wearer comfort, climate-optimizes and breathable materials
- Virtually seam-free construction made from high-tech materials to eliminate pressure points
- Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
- Soft, padding on collar and tongue
- For light applications
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6599.1
- Standard EN ISO 20345:2011 S1 P SRC
- Sizes: 35 to 52
Specifications
Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Textile | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S1 |