Learn More
SPEX™ SamplePrep Cryo-Blocks
Enables high-throughput cryogenic grinding in the Spex Geno/Grinder
Brand:  SPEX™ SamplePrep 2666
Additional Details : Weight : 1.05092kg
 View more versions of this product
Product Code. 15370462
  • 6084.77 NOK / Pack of 2
Estimated Shipment
26-04-2019
Add to basket
Description
Description
  • Chilled in the 2600 Cryo-Station and then used for high-throughput cryogenic grinding
  • Maintain samples at liquid nitrogen temperatures during tissue homogenization and cell lysis
  • Preserve pesticides, RNA, and proteins for heat sensitive extraction methods
  • Increase yields by grinding samples at sub-zero temperatures
  • Durable aluminum construction
Model-Specific Features
2660
  • Holds six 2251-PC 15mL short polycarbonate vials
  • Each pair holds up to 12 vials for rapid and efficient sample preparation
  • Used two at a time in the standard clamp assembly to keep it in balance
2661
  • Holds fifteen 2252-PC-30 15mL polycarbonate cryovials
  • Each pair holds up to 30 Cryovials for rapid and efficient sample preparation
  • Must be used two at a time in the large capacity clamp assembly to keep it in balance
2662
  • Holds twenty-four 2241-PC 5mL polycarbonate vials (as used in 2240-PC 5mL polycarbonate vial set)
  • Each pair holds up to 48 vials for rapid and efficient sample preparation
  • Used two at a time in the Standard Clamp Assembly to keep it in balance
2663
  • Holds twenty-four 2241-PE 5mL polyethylene vials (as used in 2240-PE 5mL polyethylene vial set)
  • Each pair holds up to 48 vials for rapid and efficient sample preparation
  • Used two at a time in the Standard Clamp Assembly to keep it in balance
2664
  • Holds six 2252-PC-48 50mL polycarbonate cryovials or six 2252-PC-48 50mL polyethylene cryovials
  • Each pair holds up to 12 Cryovials for rapid and efficient sample preparation
  • Must be used two at a time in the large capacity clamp assembly to keep it in balance
2665
  • Holds 96 standard (0.6mL) microcentrifuge or PCR tubes
  • Each pair holds up to 192 tubes for rapid and efficient sample preparation
  • Used two at a time in the standard clamp assembly to keep it in balance
2666
  • Holds 48 standard (1.5 - 2.0mL) microcentrifuge or PRC tubes
  • Each pair holds up to 96 tubes for rapid and efficient sample preparation
  • Used two at a time in the standard clamp assembly to keep it in balance
Specifications
Specifications
Ideal for high throughput cryogenic grinding and to preserve pesticides, RNA, Proteins and other temperature sensitive samples
Aluminum
Cryo Block
48 Standard 1.5 to 2mL microcentrifuge or PRC tubes, each pair holds up to 96 tubes
Spex Geno/Grinder
Videos