Learn More
Cole-Parmer™ Plastic Vials for High-Energy Mixer/Mill™ Grinders
Use these Plastic Vials with high-energy Mixer/Mill™ grinders to pulverize and mix samples.
Brand: Cole-Parmer™ 8002-SSP
Additional Details : Weight : 5.00000kg
Description
- Suitable for use with Cole-Parmer™ Mixer/Mill™ High-Energy Ball Mills.
- Order methacrylate balls to use with disposable plastic vials.
Specifications
Grinding Vial | |
54 mm | |
8000 D and 8000 M Mixer/Mills | |
67 mm |
Screw-on Cap | |
Ideal for Pulverizing Soft, Brittle Materials and for Mixing Liquids and Powders, Slurry Grinding | |
2.5 in. | |
Polystyrene |