Learn More
Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits
For smaller dialysis experiments or evaluating dialysis tubing for dialysis application
Brand: Spectrum™ 132 684T1
Additional Details : Weight : 0.10000kg
Includes:
- 1m or 5m of Spectra/Por 2 or 4 dialysis tubing
- One Standard Spectra/Por Closure (for buoyance)
- One weighted Spectra/Por Closure (for orientation)
- Two adhesive tubing openers
Description
- Hydrophilic, symmetric porosity
- Ensures higher permeability and basic dialysis
- Available as dry with glycerin (soak in water)
Specifications
12 to 14 kDa | |
76 mm | |
46 mL/cm vol./length | |
1 m Tubing/Kit |