Learn More
Lovibond™ Spectroquant™ Cadmium Tube Test, 0.025 to 0.75mg/L
For use with Lovibond test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 420750
Additional Details : CAS Number : 9002-93-1 Weight : 0.30000kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Description
- Range: 0.025 to 0.75mg/L
- Parameter: Cadmium
- Method: Cadion
- For use in test kits, colorimeters and spectrophotometers
- Case of 25
Specifications
Test kits, colorimeters and spectrophotometers | |
0.025 to 0.75mg/L | |
Cadmium Tube Test |
25 | |
Cadion |
Safety and Handling
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : triton-X-100, disodium tetraborate