missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Antibody (124413) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB942JF525
This item is not returnable.
View return policy
Description
SPARC Monoclonal antibody specifically detects SPARC in Mouse samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| SPARC | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| Mouse myeloma cell line NS0-derived recombinant mouse SPARC/Osteonectin, Ala18-Ile302, Accession # P07214 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 124413 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.5 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
| 6678 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction