missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | SPAG7 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885884
|
Proteintech
16294-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16875884
|
Proteintech
16294-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SPAG7 gene ontology annotations related to this gene include nucleic acid binding.Specifications
SPAG7 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O75391, Q7TNE3 | |
216873, 303260, 9552 | |
SPAG7 Fusion Protein Ag9471 | |
Primary | |
-20°C | |
SPAG7 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ACRP, FSA 1, SPAG7, sperm associated antigen 7 | |
SPAG7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |