Learn More
H-B Frio-Temp SP Scienceware™ Calibrated Electronic Verification Thermometers, Calibrated for General Applications
Shop All SP Scienceware Products
Click to view available options
Bottle Size:
10 mL
30 mL
Bottle Fill:
Glass Bottle/Glass Beads
Plastic Bottle (2)/Glass Beads
Plastic Bottle (2)/Organic Liquid
Plastic Bottle/Glass Beads
Length (Metric) Cable:
1 m
1.5 m
3 m
Unit Size:
Each
Description
Includes
Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Requires
AAA battery
- Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
- All models include accredited calibration documentation that meets the requirements of ISO 17025
- Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No. 2448.01), against equipment whose calibration is traceable to NIST
- Individually serialized, CE marked
Certifications
CE marked
Compliance
Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized.
Specifications
Specifications
| Bottle Fill | Glass Bottle/Glass Beads |
| Bottle Size | 30 mL |
| Calibration Points | 0°, 100°C |
| Certifications/Compliance | Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE marked. |
| For Use With (Application) | Temperature Calibration and Verification |
| Height (English) | 4.3 in. |
| Height (Metric) | 110 mm |
| Includes | Multi-language instructions (English, French, German, Italian, Portuguese and Spanish) |
| Length (English) Cable | 9.8 ft. |
| Length (English) Probe | 3.1 in. |
| Show More |