Learn More
Sodium Thiosulfate Solution 0.5M (0.5N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1AMP
6AMP
Specifications
Boiling Point | 100°C |
Melting Point | 0°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 1AMP |
Packaging | Ampoule |
Vapor Pressure | 14mmHg |