missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide NF/FCC Pellet, Macron Fine Chemicals™

Catalog Number 15588424
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
Quantity 500 g
Formula Weight 40
Grade FCC/NF
Packaging POLYSTORMOR
Physical Form Pellet
Chemical Name or Material Sodium Hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'