Learn More
Sodium hydrogencyanamide, 98%, Thermo Scientific Chemicals
Catalog Number p-4258717
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
17292-62-5
CHN2Na
64.02
MFCD00066500
MBEGFNBBAVRKLK-UHFFFAOYSA-N
sodium hydrogencyanamide, sodium acid cyanamide, sodium cyanamide, sodium cyanamidate, sodium cyanoazanide, cyanamide, monosodium salt, monosodium cyanamide, sodium hydrogen cyanamide, cyanamide, sodium salt 1:1, cyanamide monosodium salt
4293993
[Na+].[NH-]C#N
17292-62-5
CHN2Na
64.02
MFCD00066500
MBEGFNBBAVRKLK-UHFFFAOYSA-N
sodium hydrogencyanamide, sodium acid cyanamide, sodium cyanamide, sodium cyanamidate, sodium cyanoazanide, cyanamide, monosodium salt, monosodium cyanamide, sodium hydrogen cyanamide, cyanamide, sodium salt 1:1, cyanamide monosodium salt
4293993
[Na+].[NH-]C#N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point >360.0°C
Color Tan to White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98%
Solubility Information Solubility in water: 800g/L (20°C)
IUPAC Name sodium cyanoazanide
Formula Weight 64.02
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Sodium hydrogencyanamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium hydrogencyanamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.