missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, >99%, MP Biomedicals™

Catalog Number p-4404527
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
This item is not returnable. View return policy
151-21-3
C12H25NaO4S
288.38
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
3423265
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
This item is not returnable. View return policy
151-21-3
C12H25NaO4S
288.38
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
3423265
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Color White
Density >1.1 at 20°C
Quantity 10 g
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Percent Purity >99%
Physical Form Granular Powder
Chemical Name or Material Sodium dodecyl sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'