missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride USP/FCC Granular, Macron Fine Chemicals™
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly
Specifications
Physical Form | Granules |
Quantity | 2.5 kg |
IUPAC Name | sodium;chloride |
Formula Weight | 58.44 |
Grade | FCC/USP |
Packaging | Poly |
Chemical Name or Material | Sodium Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'