missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073511
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7775-09-9
ClNaO3
106.44
3479
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
516902
CHEBI:65242
sodium chlorate
[Na+].[O-][Cl](=O)=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7775-09-9
ClNaO3
106.44
3479
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
516902
CHEBI:65242
sodium chlorate
[Na+].[O-][Cl](=O)=O
Melting Point 248°C
Color White
Physical Form Solid
Quantity 500 g
Formula Weight 106.44g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium chlorate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Oxidising solid Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'