missing translation for 'onlineSavingsMsg'
Learn More

Sodium borodeuteride, 99% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7050061
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
15681-89-7
BH4Na
37.83
MFCD00003519
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride
23673181
sodium boranuide
[BH4-].[Na+]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
15681-89-7
BH4Na
37.83
MFCD00003519
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride
23673181
sodium boranuide
[BH4-].[Na+]

Sodium borodeuteride is used to prepare alfa, alfa-dideutero-beta-amino acids. It is also used to prepare amine-stabilized, deuterated borane-tetrhydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Color White
Sensitivity Moisture
Quantity 1 g
pH 11
Melting Point ∼400°C (decomposition)
Linear Formula NaBD4
UN Number UN1426
Solubility Information Soluble in liquid ammonia,amines and pyridine.
Formula Weight 41.88
Percent Purity 99%
Chemical Name or Material Sodium borodeuteride, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'