Learn More
Sodium Benzoate-d5, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Sodium Benzoate-d5 |
| CAS | 62790-26-5 |
| Molecular Formula | C7 D5 O2 . Na |
| Quantity | 1 g |
| Synonym | Benzoic-d5 acid, sodium salt (9 CI), Sodium benzoate-d5, Benzoic-2,3,4,5,6-d5 acid sodium salt, 2,3,4,5,6-Pentadeuteriobenzoic acid sodium salt, Benzoic Acid-D5 (phenyl-D5) Sodium Salt |
| InChI Formula | InChI=1 S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5 H,(H,8,9);/q;+1/p-1/i1D,2 D,3 D,4 D,5 D; |
| SMILES | [Na+].[2 H]c1c([2 H])c([2 H])c(C(=O)[O-])c([2 H])c1[2 H] |
| IUPAC Name | sodium;2,3,4,5,6-pentadeuteriobenzoate |
| Molecular Weight (g/mol) | 149.134 |
| Formula Weight | 149.05 g/mol |
| Show More |