missing translation for 'onlineSavingsMsg'
Learn More

Sodium aurothiomalate(I), 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050930
Click to view available options
Quantity:
1 g
5 g
12244-57-4
C4H5AuNa2O5S
408.09
MFCD00064304,MFCD00064304
YLQOAPBVYJCTPW-UHFFFAOYNA-K
gold sodium thiomalate
133108869
gold;sodium;2-sulfanylbutanedioic acid
O.[Na+].[Na+].[Au+].[O-]C(=O)CC([S-])C([O-])=O
12244-57-4
C4H5AuNa2O5S
408.09
MFCD00064304,MFCD00064304
YLQOAPBVYJCTPW-UHFFFAOYNA-K
gold sodium thiomalate
133108869
gold;sodium;2-sulfanylbutanedioic acid
O.[Na+].[Na+].[Au+].[O-]C(=O)CC([S-])C([O-])=O

Sodium aurothiomalate(I) is useful for high mobility group box chromosomal protein 1 western blotting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Sulphurous
Quantity 1 g
Merck Index 14,4518
Solubility Information Very soluble in water; Practically insoluble in alcohol, ether
Formula Weight 390.08
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Sodium aurothiomalate(I)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'