missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, Free-flowing, ACS reagent, anhydrous, ≥99.0%, Honeywell Fluka™

Catalog Number p-7111981
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium acetate
[Na+].CC([O-])=O
This item is not returnable. View return policy
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium acetate
[Na+].CC([O-])=O

Free-flowing, ACS reagent, anhydrous,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point >300°C (decomposition)(literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Physical Form Anhydrous
Quantity 100 g
Linear Formula CH3COONa
Beilstein 3595639
Formula Weight 82.03
Percent Purity >99%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.45 g/cm3
Assay HClO4-Titr.
Chemical Name or Material Sodium acetate, Free-flowing, ACS reagent, anhydrous, ≥99.0%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'