missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, 3M aq. soln., pH 7.0, autoclaved, Thermo Scientific Chemicals

Catalog Number p-7041973
Click to view available options
Quantity:
125 mL
250 mL
This item is not returnable. View return policy
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium;acetate
[Na+].CC([O-])=O
This item is not returnable. View return policy
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium;acetate
[Na+].CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Linear Formula NaOOCCH3
Concentration 3M aq. soln.
Formula Weight 82.03
Odor Odorless
pH 7.0
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Sodium acetate, autoclaved

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'