missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-dibenzofuransulfonate hydrate, 98+%, Thermo Scientific™

Catalog Number p-7046167
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
94600-19-8
C12H7NaO4S
270.23
MFCD00046899
MITIQPHWKOQOAR-UHFFFAOYSA-M
sodium 2-dibenzofuransulphonate hydrate, sodium 2-dibenzofuransulfonate hydrate, sodium dibenzo b,d furan-2-sulfonate-water 1/1/1, sodium 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2 7 ,3,5,9,11-hexaene-4-sulfonate hydrate, sodium 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonate hydrate, sodium 8-oxatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonate hydrate
57356836
sodium;dibenzofuran-2-sulfonate;hydrate
[Na+].[O-]S(=O)(=O)C1=CC2=C(OC3=C2C=CC=C3)C=C1
This item is not returnable. View return policy
94600-19-8
C12H7NaO4S
270.23
MFCD00046899
MITIQPHWKOQOAR-UHFFFAOYSA-M
sodium 2-dibenzofuransulphonate hydrate, sodium 2-dibenzofuransulfonate hydrate, sodium dibenzo b,d furan-2-sulfonate-water 1/1/1, sodium 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2 7 ,3,5,9,11-hexaene-4-sulfonate hydrate, sodium 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonate hydrate, sodium 8-oxatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonate hydrate
57356836
sodium;dibenzofuran-2-sulfonate;hydrate
[Na+].[O-]S(=O)(=O)C1=CC2=C(OC3=C2C=CC=C3)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 1 g
Beilstein 3795215
Formula Weight 270.24 (Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Sodium 2-dibenzofuransulfonate hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'