missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 2-bromoethanesulfonate, 98%, Thermo Scientific Chemicals
258.01 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Sodium 2-bromoethanesulfonate(BES) was used to investigate the effect of BES on inhibition of bacterial growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium 2-bromoethanesulfonate(BES) was used to investigate the effect of BES on inhibition of bacterial growth.
Solubility
Soluble in water and methanol.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Sodium 2-bromoethanesulfonate(BES) was used to investigate the effect of BES on inhibition of bacterial growth.
Solubility
Soluble in water and methanol.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Melting Point | ∼284°C (decomposition) |
Quantity | 25 g |
Beilstein | 4590828 |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 211.02 |
Percent Purity | 98% |
Chemical Name or Material | Sodium 2-bromoethanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'