Learn More
Lenz Laborglasintrumente Sockets with Special Tube Dimensions
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01101005
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10050331
  • 56.35 NOK / Each
Estimated Shipment: 20-07-2022
Description
  • Sockets with special tube dimensions
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Specifications
1 Ea.
2 mm
210 mm
NS 29/32
33 mm
Duran Glass